Learn More
Haematologic Technologies Inc FACTOR XIA SUBSTRATE 1MG
Supplier:  Haematologic Technologies Inc SN13A
Encompass_Preferred
Fluorogenic substrate, Contains the fluorescent reporter group 6-amino-1-naphthalene-sulfonamide (ANSN) for monitoring enzyme activity of the serine protease FACTOR XIa, D-LPR-ANSNH-C3H7 • 2 HCl, 1 milligram, RUO, Supplied in DMSO, store at -20 Celsius.
Catalog No. 50-883-408