Learn More
BINDER™ Factory Calibration Certificate Extension
SDP
Catalog No. 08111586 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers B 28
Catalog No. For Use With (Equipment)
08-111-586 BINDER Chambers B 28
1 options
Catalog No. 08-111-586 Supplier BINDER™ Supplier No. 80120022
Add to Cart
Add to Cart
For Use With: BINDER units B 28
  • For extending the measurement in center of chamber to include another test temperature
  • For use with all ED, FD, FED, FP, M, MK, BD, BF, KB, KBW models and FDL 115, MDL 115
Specifications
Description Calibration certificate, expanded - for temperature; for extending the measurement in center of chamber to include another test temperature
For Use With (Equipment) BINDER Chambers B 28
Type Calibration Certificate