Learn More
Thermco Products Inc FACTORY CERT THERMOMETER
Catalog No. NC0999156
Encompass_Preferred
ASTM 114C thermometer (red spirit) certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.
Catalog No. NC0999156 Supplier Thermco Products Inc Supplier No. ACC114CFC
ASTM 114C thermometer (red spirit) certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.