Learn More
Enzo Life Sciences FADD monoclonal antibody (1F7) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM212E
Encompass_Preferred
Clone: 1F7. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human FADD. UniProt: Q13158. Species Reactivity: Human, Mouse. Applications: WB. Recommended dilutions: Western Blot (1µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-658-723