Learn More
Aviva Systems Biology Corp FADD Antibody - C-terminal region (ARP30294_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30294_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fadd. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse FADD. Alternate names: Mort1/FADDApplicationWB. Species reactivity: Mouse
Catalog No. 50-014-54148