Learn More
AdipoGen anti-FADD (human), mAb (1C4) (Biotin) (100 µg)
SDP
Supplier:  AdipoGen AG20B0080BC100
Encompass_Preferred
Monoclonal Antibody. Recognizes the C-terminus of human FADD only in its unphosphorylated form. Application: ELISA,IP,WB. Clone: 1C4. Isotype: Mouse IgG1. Liquid. In PBS and 0.02% sodium azide. FADD (FAS-associated death domain protein) is an apoptotic adaptor molecule that recruits caspase-8 or caspase-10 to the activated Fas (CD95) or TNFR-1 receptors. Procaspase-8 belongs to the family of caspases. Binding of FasL to Fas leads to formation of a receptor complex at the cellular membrane, which was named DISC. The DISC structure provides a platform for the oligomerization of procaspase-8 that allows two procaspase-8 homodimers to be in the close proximity leading to the initial activation of procaspase-8. FADD is also involved in interferon-mediated antiviral immune response and functions in the positive regulation of interferon signaling. FADD also plays a role in embryonic development and the cell cycle reentry of T cells.
Catalog No. 50-205-2995