Learn More
Cayman Chemical FAD (sodium salt), Purity: ≥98%, CAS: 84366-81-4, 500 mg
SDP
Catalog No. 502987967
Encompass_Preferred
A flavin dinucleotide; functions as a coenzyme that facilitates the transfer of electrons by flavoenzymes in oxidation-reduction reactions; reduces UV-B-induced death of human corneal epithelial cells in vitro
Catalog No. 50-298-7967 Supplier Cayman Chemical Supplier No. 23386500 MG
A flavin dinucleotide; functions as a coenzyme that facilitates the transfer of electrons by flavoenzymes in oxidation-reduction reactions; reduces UV-B-induced death of human corneal epithelial cells in vitro