Learn More
Medchemexpress LLC HY-101212 10mg , Fadraciclib CAS:1070790-89-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10121210MG
Encompass_Preferred
HY-101212 10mg Fadraciclib CAS: 1070790-89-4 Fadraciclib (CYC065) is a second-generation, orally available ATP-competitive inhibitor of CDK2/CDK9 kinases with IC50s of 5 and 26 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9423