Learn More
Medchemexpress LLC Fadrozole hydrochloride | 102676-31-3 | 99.9% | 259.73 | 5 MG
SDP
Catalog No. 5000364361
Encompass_Preferred
Fadrozole hydrochloride is a potent, selective, and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM. This product is for research use only and is not intended for patient treatment.
  • Inhibits aromatase with an IC50 of 6.4 nM
  • Inhibits estrogen production in hamster ovarian slices with an IC50 of 0.03 μM
  • Prevents the development of both benign and malignant spontaneous mammary neoplasms in female Sprague-Dawley rats
  • Slows the spontaneous development of pituitary pars distalis adenomas and reduces the incidence of spontaneous hepatocellular tumors
  • Reduces parasite burden in male and female mice and recovers specific cellular immune response in male mice
Catalog No. 50-003-64361 Supplier Medchemexpress LLC Supplier No. 5MGHY14247B
Fadrozole hydrochloride is a potent, selective, and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM. This product is for research use only and is not intended for patient treatment.
  • Inhibits aromatase with an IC50 of 6.4 nM
  • Inhibits estrogen production in hamster ovarian slices with an IC50 of 0.03 μM
  • Prevents the development of both benign and malignant spontaneous mammary neoplasms in female Sprague-Dawley rats
  • Slows the spontaneous development of pituitary pars distalis adenomas and reduces the incidence of spontaneous hepatocellular tumors
  • Reduces parasite burden in male and female mice and recovers specific cellular immune response in male mice