Learn More
Medchemexpress LLC Fadrozole hydrochloride | 102676-31-3 | 99.9% | C14H14ClN3 | 1 ML
SDP
Catalog No. 5000354086
Encompass_Preferred
Fadrozole hydrochloride (CGS 16949A) is a potent, selective, and nonsteroidal aromatase inhibitor that effectively inhibits estrogen production. It is intended for research use only and not for patient use.
  • Potent and selective aromatase inhibitor with an IC50 of 6.4 nM.
  • Inhibits estrogen production in hamster ovarian slices with an IC50 of 0.03 μM.
  • Can suppress the synthesis of other cytochrome P-450 dependent steroids at higher doses.
  • Prevents the development of spontaneous mammary neoplasms in female rats.
  • Slows the development of pituitary pars distalis adenomas.
  • Reduces the incidence of spontaneous hepatocellular tumors.
  • Reduces parasite burden in male and female mice by 70%.
Catalog No. 50-003-54086 Supplier Medchemexpress LLC Supplier No. HY1424710MM/1ML
Fadrozole hydrochloride (CGS 16949A) is a potent, selective, and nonsteroidal aromatase inhibitor that effectively inhibits estrogen production. It is intended for research use only and not for patient use.
  • Potent and selective aromatase inhibitor with an IC50 of 6.4 nM.
  • Inhibits estrogen production in hamster ovarian slices with an IC50 of 0.03 μM.
  • Can suppress the synthesis of other cytochrome P-450 dependent steroids at higher doses.
  • Prevents the development of spontaneous mammary neoplasms in female rats.
  • Slows the development of pituitary pars distalis adenomas.
  • Reduces the incidence of spontaneous hepatocellular tumors.
  • Reduces parasite burden in male and female mice by 70%.