Learn More
Medchemexpress LLC Fadrozole hydrochloride | 102676-31-3 | 99.9% | C14H14ClN3 | 100 MG
SDP
Catalog No. 5000353637
Encompass_Preferred
Fadrozole hydrochloride (CGS 16949A) is a potent, selective, and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM. Intended for research use only, this compound effectively inhibits estrogen and progesterone production. It has demonstrated efficacy in preventing the development of various spontaneous neoplasms and reducing parasite burden in animal models.
  • Potent, selective, and nonsteroidal aromatase inhibitor
  • Inhibits estrogen production in hamster ovarian slices
  • Inhibits progesterone production
  • Prevents development of benign and malignant spontaneous mammary neoplasms
  • Reduces incidence of spontaneous hepatocellular tumors
  • Reduces parasite burden in male and female mice
Catalog No. 50-003-53637 Supplier Medchemexpress LLC Supplier No. HY14247100MG
Fadrozole hydrochloride (CGS 16949A) is a potent, selective, and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM. Intended for research use only, this compound effectively inhibits estrogen and progesterone production. It has demonstrated efficacy in preventing the development of various spontaneous neoplasms and reducing parasite burden in animal models.
  • Potent, selective, and nonsteroidal aromatase inhibitor
  • Inhibits estrogen production in hamster ovarian slices
  • Inhibits progesterone production
  • Prevents development of benign and malignant spontaneous mammary neoplasms
  • Reduces incidence of spontaneous hepatocellular tumors
  • Reduces parasite burden in male and female mice