Learn More
Medchemexpress LLC Fadrozole hydrochloride | 102676-31-3 | MFCD02313480 | 99.9% | C14H14ClN3 | 10MG
SDP
Catalog No. 5000261999
Encompass_Preferred
Fadrozole hydrochloride is a potent, selective, nonsteroidal aromatase inhibitor used in biochemical and pharmacological research to inhibit estrogen biosynthesis. It is supplied as the hydrochloride salt with high reported purity and is commonly used in in vitro assays to evaluate aromatase activity and inhibitor potency. The reported IC50 for aromatase inhibition is 6.4 nM.
  • Potent aromatase inhibition (IC50 6.4 nM).
  • High purity (>99.9%).
  • Supplied as a hydrochloride salt for improved handling and solubility.
  • Suitable for in vitro biochemical and pharmacological assays.
  • Molecular formula C14H14ClN3.
Catalog No. 50-002-61999 Supplier Medchemexpress LLC Supplier No. HY1424710MG
Fadrozole hydrochloride is a potent, selective, nonsteroidal aromatase inhibitor used in biochemical and pharmacological research to inhibit estrogen biosynthesis. It is supplied as the hydrochloride salt with high reported purity and is commonly used in in vitro assays to evaluate aromatase activity and inhibitor potency. The reported IC50 for aromatase inhibition is 6.4 nM.
  • Potent aromatase inhibition (IC50 6.4 nM).
  • High purity (>99.9%).
  • Supplied as a hydrochloride salt for improved handling and solubility.
  • Suitable for in vitro biochemical and pharmacological assays.
  • Molecular formula C14H14ClN3.