Learn More
Medchemexpress LLC HY-14247 5mg Medchemexpress, Fadrozole hydrochloride CAS:102676-31-3 Purity:>98%
SDP
Catalog No. 502081576
Encompass_Preferred
Medchemexpress, HY-14247 5mg Fadrozole hydrochloride CAS:102676-31-3 Fadrozole hydrochloride is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1576 Supplier Medchemexpress LLC Supplier No. HY142475MG
Medchemexpress, HY-14247 5mg Fadrozole hydrochloride CAS:102676-31-3 Fadrozole hydrochloride is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.