Learn More
Medchemexpress LLC Fadrozole hydrochloride (CGS 16949A) | 102676-31-3 | 99.9% | C14H14ClN3 | 50 MG
SDP
Catalog No. 5000354087
Encompass_Preferred
Fadrozole hydrochloride (CGS 16949A) is a potent, selective, and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM. It inhibits estrogen production in hamster ovarian slices with an IC50 of 0.03 μM and progesterone production with an IC50 of 120 μM. Higher doses can suppress the synthesis of other cytochrome P-450 dependent steroids to varying degrees.
  • Potent and selective aromatase inhibitor.
  • Prevents development of benign and malignant spontaneous mammary neoplasms.
  • Slows development of pituitary pars distalis adenomas and reduces incidence of spontaneous hepatocellular tumors.
  • Reduces parasite burden in male and female mice by 70%.
  • Augments Interleukin-6 (IL-6) serum levels and production by splenocytes.
  • Nonsteroidal option for aromatase inhibition.
Catalog No. 50-003-54087 Supplier Medchemexpress LLC Supplier No. HY1424750MG
Fadrozole hydrochloride (CGS 16949A) is a potent, selective, and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM. It inhibits estrogen production in hamster ovarian slices with an IC50 of 0.03 μM and progesterone production with an IC50 of 120 μM. Higher doses can suppress the synthesis of other cytochrome P-450 dependent steroids to varying degrees.
  • Potent and selective aromatase inhibitor.
  • Prevents development of benign and malignant spontaneous mammary neoplasms.
  • Slows development of pituitary pars distalis adenomas and reduces incidence of spontaneous hepatocellular tumors.
  • Reduces parasite burden in male and female mice by 70%.
  • Augments Interleukin-6 (IL-6) serum levels and production by splenocytes.
  • Nonsteroidal option for aromatase inhibition.