Learn More
Medchemexpress LLC Fadrozole hydrochloride hemihydrate | 176702-70-8 | 99.6% | 268.74 | 25 MG
SDP
Catalog No. 5000365771
Encompass_Preferred
Fadrozole hydrochloride hemihydrate is an orally active, potent, selective, and nonsteroidal aromatase inhibitor, with an IC50 of 6.4 nM. It inhibits the production of estrogen and progesterone, with IC50 values of 0.03 and 120 μM, respectively. This compound shows prevention of spontaneous tumors and can be used for research on estrogen-dependent diseases and cancer.
  • Orally active
  • Potent, selective, and nonsteroidal aromatase inhibitor (IC50 of 6.4 nM)
  • Inhibits estrogen production (IC50 of 0.03 μM)
  • Inhibits progesterone production (IC50 of 120 μM)
  • Prevents spontaneous tumors
  • Useful for research of estrogen-dependent diseases and cancer
Catalog No. 50-003-65771 Supplier Medchemexpress LLC Supplier No. 25MGHY14247B
Fadrozole hydrochloride hemihydrate is an orally active, potent, selective, and nonsteroidal aromatase inhibitor, with an IC50 of 6.4 nM. It inhibits the production of estrogen and progesterone, with IC50 values of 0.03 and 120 μM, respectively. This compound shows prevention of spontaneous tumors and can be used for research on estrogen-dependent diseases and cancer.
  • Orally active
  • Potent, selective, and nonsteroidal aromatase inhibitor (IC50 of 6.4 nM)
  • Inhibits estrogen production (IC50 of 0.03 μM)
  • Inhibits progesterone production (IC50 of 120 μM)
  • Prevents spontaneous tumors
  • Useful for research of estrogen-dependent diseases and cancer