Learn More
Aviva Systems Biology Corp FADS1 Antibody - N-terminal region (ARP42383_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42383_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FADS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FADS1. Alternate names: D5D, FADS6, FADSD5, FLJ38956, FLJ90273, LLCDL1, TU12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58207