Learn More
ATCC Faecalibacterium prausnitzii; Strain: VPI C13-51
Catalog No. 502382227
Encompass
Faecalibacterium prausnitzii strain VPI C13-51 was isolated from human feces in Blacksburg, Virginia. This whole-genome sequenced bacterial type strain is a strict anaerobe.
Catalog No. 50-238-2227 Supplier ATCC Supplier No. 27768
Faecalibacterium prausnitzii strain VPI C13-51 was isolated from human feces in Blacksburg, Virginia. This whole-genome sequenced bacterial type strain is a strict anaerobe.