Learn More
Aviva Systems Biology Corp FAF1 Antibody - C-terminal region (ARP61635_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61635_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAF1. It was validated on Western Blot. . Alternate names: CGI-03, FLJ37524, HFAF1s, UBXD12, UBXN3A, hFAF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-287-427