Learn More
Aviva Systems Biology Corp FAHD2A Antibody - N-terminal region (ARP62545_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62545_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAHD2A. It was validated on Western Blot. . Alternate names: CGI-105, MGC131995ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-287-710