Learn More
LGC GENOMICS LLC (Lucigen) FailSafe Enzyme Mix Only, 100 U @2.5 U/LPCR Enzyme Mix, individual component of the FailSafe PreMix Selection Kit for PCR optimization, and the bundled post - optimization FailSafe PCR System.
Catalog No. NC9366063
Encompass
The FailSafe™ PCR System provides dependable, consistent high-fidelity PCR results for every DNA template, regardless of its source or sequence. The FailSafe PCR System will faithfully amplify your template every time.We promise!
This item is not returnable. View return policy
Catalog No. NC9366063 Supplier LGC GENOMICS LLC Supplier No. FSE51100
The FailSafe™ PCR System provides dependable, consistent high-fidelity PCR results for every DNA template, regardless of its source or sequence. The FailSafe PCR System will faithfully amplify your template every time.We promise!