Learn More
LGC GENOMICS LLC (Lucigen) FAILSAFE PCR PREMIX C 2.5MLTwelve 2X PreMixes, individual components of the FailSafe PreMix Selection Kit for PCR optimization, and the bundled post - optimization FailSafe PCR System.
Catalog No. NC2162055
Encompass
Trusted: The same great performance as the FailSafe PCR System, but available as individual componentsFlexible: Purchase any combination of FailSafe 2X PreMixesThis listing is only for reordering of individual FailSafe PCR 2X PreMixes.Before purchasing these separate components, we strongly recommend first optimizing your PCR conditions using the FailSafe PCR PreMix Selection Kit (Item ID. FS99060) in order to know which components to purchase.
This item is not returnable. View return policy
Catalog No. NC2162055 Supplier LGC GENOMICS LLC Supplier No. FSP995C
Trusted: The same great performance as the FailSafe PCR System, but available as individual componentsFlexible: Purchase any combination of FailSafe 2X PreMixesThis listing is only for reordering of individual FailSafe PCR 2X PreMixes.Before purchasing these separate components, we strongly recommend first optimizing your PCR conditions using the FailSafe PCR PreMix Selection Kit (Item ID. FS99060) in order to know which components to purchase.