Learn More
Aviva Systems Biology Corp FAIM Antibody - middle region (ARP56238_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56238_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAIM. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAIM. Alternate names: FAIM1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61777