Learn More
MSC Fairbanks Caster Wheel, Polyolefin, 6 in Dia, Plain Bearing
Supplier: MSC 65366239
This Fairbanks caster wheel features a durable polyolefin tread and a plastic core, designed for various industrial and commercial applications. With a 6 in diameter and 2 in width, it is equipped with a plain bearing for smooth operation. Its non-marking property and wide temperature range of -20 F to 230 F make it suitable for environments such as food processing, warehousing, office operations, and manufacturing.
- Polyolefin tread and plastic core for durability.
- 6 in diameter and 2 in width.
- Plain bearing for smooth operation.
- Non-marking.
- Suitable for food processing, warehousing, office operations, and manufacturing.
- Operates in temperatures from -20 F to 230 F.