Learn More
MSC Fairbanks Rigid Top Plate Caster, Polyurethane, 6 in Wheel Dia, 1,000 lb Capacity
Catalog No. 5000510555
Encompass
This Rigid Top Plate Caster features a polyurethane wheel with a 6-inch diameter and 2-inch width, molded onto a cast iron core. It offers a 1,000 lb capacity and an overall height of 7-1/4 inches, suitable for industrial and institutional applications.
  • One-piece swivel caster horn construction
  • Hardened top plate and fork
  • Nylon grease seal ring on the upper ball raceway
  • Orbit formed king pin
  • Epoxy powder coated finish
  • Plate size 4 x 4-1/2 in
  • Roller bearing type
Catalog No. 50-005-10555 Supplier MSC Supplier No. 65365900
This Rigid Top Plate Caster features a polyurethane wheel with a 6-inch diameter and 2-inch width, molded onto a cast iron core. It offers a 1,000 lb capacity and an overall height of 7-1/4 inches, suitable for industrial and institutional applications.
  • One-piece swivel caster horn construction
  • Hardened top plate and fork
  • Nylon grease seal ring on the upper ball raceway
  • Orbit formed king pin
  • Epoxy powder coated finish
  • Plate size 4 x 4-1/2 in
  • Roller bearing type