Learn More
MSC Fairbanks Rigid Top Plate Caster, Rubber, 4 in Wheel, 200 lb
Catalog No. 5000516979
Encompass
This rigid top plate caster features durable Lockweld construction and a zinc finish, designed for medium-duty applications. It includes a 4 inch rubber wheel with a 200 lb load capacity, making it suitable for various industrial and commercial uses.
  • Lockweld construction for enhanced durability
  • Plate mount for secure attachment
  • 4 inch rubber wheel for smooth operation
  • 200 lb load capacity for medium-duty applications
  • Roller bearing for efficient movement
  • Stamp formed construction for robustness
  • Zinc finish for corrosion resistance
  • Large secondary raceway provides greater swiveling, efficiency, and shock absorption
  • Double ball race swivels for improved maneuverability
  • Rubber molded on cast iron for reliability
Catalog No. 50-005-16979 Supplier MSC Supplier No. 01357144
This rigid top plate caster features durable Lockweld construction and a zinc finish, designed for medium-duty applications. It includes a 4 inch rubber wheel with a 200 lb load capacity, making it suitable for various industrial and commercial uses.
  • Lockweld construction for enhanced durability
  • Plate mount for secure attachment
  • 4 inch rubber wheel for smooth operation
  • 200 lb load capacity for medium-duty applications
  • Roller bearing for efficient movement
  • Stamp formed construction for robustness
  • Zinc finish for corrosion resistance
  • Large secondary raceway provides greater swiveling, efficiency, and shock absorption
  • Double ball race swivels for improved maneuverability
  • Rubber molded on cast iron for reliability