Learn More
MSC Fairbanks Swivel Caster, Polyurethane, 5 inch, 700 lb Capacity
Catalog No. 5000512127
Encompass
This swivel top plate caster features a 5-inch diameter polyurethane wheel, a 2-inch wheel width, and a robust 700 lb load capacity. It is designed for industrial and institutional applications, offering durability and smooth operation with its kingpin swivel construction and epoxy powder-coated finish.
  • One-piece swivel caster horn construction
  • Hardened top plate and fork for enhanced durability
  • Nylon grease seal ring on the upper ball raceway for smooth operation
  • Orbit formed king pin ensures reliable swivel action
  • Wheel molded onto a cast iron core for strength
  • Roller bearings for efficient movement
  • Sealed design protects components from contaminants
  • Epoxy powder coated finish for corrosion resistance
Catalog No. 50-005-12127 Supplier MSC Supplier No. 65365710
This swivel top plate caster features a 5-inch diameter polyurethane wheel, a 2-inch wheel width, and a robust 700 lb load capacity. It is designed for industrial and institutional applications, offering durability and smooth operation with its kingpin swivel construction and epoxy powder-coated finish.
  • One-piece swivel caster horn construction
  • Hardened top plate and fork for enhanced durability
  • Nylon grease seal ring on the upper ball raceway for smooth operation
  • Orbit formed king pin ensures reliable swivel action
  • Wheel molded onto a cast iron core for strength
  • Roller bearings for efficient movement
  • Sealed design protects components from contaminants
  • Epoxy powder coated finish for corrosion resistance