Learn More
MSC Fairbanks Swivel Top Plate Caster, 6 in Wheel, 1,200 lb Capacity, Semi-Steel
Catalog No. 5000510554
Encompass
This swivel top plate caster is designed for industrial and institutional applications. It features a semi-steel wheel, a 6-inch wheel diameter, and a robust 1,200 lb load capacity, with an overall height of 7-1/4 inches. The caster is constructed with a kingpin swivel and finished with an epoxy powder coating, ensuring durability and smooth operation.
  • One-piece swivel caster horn construction
  • Hardened top plate and fork
  • Nylon grease seal ring on the upper ball raceway
  • Orbit formed king pin
  • Plain bearing type
  • Sealed bearing
  • Stamp formed construction
  • Gray wheel color
  • 4 x 4-1/2 inch plate size
Catalog No. 50-005-10554 Supplier MSC Supplier No. 65365769
This swivel top plate caster is designed for industrial and institutional applications. It features a semi-steel wheel, a 6-inch wheel diameter, and a robust 1,200 lb load capacity, with an overall height of 7-1/4 inches. The caster is constructed with a kingpin swivel and finished with an epoxy powder coating, ensuring durability and smooth operation.
  • One-piece swivel caster horn construction
  • Hardened top plate and fork
  • Nylon grease seal ring on the upper ball raceway
  • Orbit formed king pin
  • Plain bearing type
  • Sealed bearing
  • Stamp formed construction
  • Gray wheel color
  • 4 x 4-1/2 inch plate size