Learn More
Electron Microscopy Sciences Fairs Microscope Graticle NG5, 19mm, Grid length 4 mm
SDP
Catalog No. 5025882
Encompass_Preferred
Designed to extend the sizing range of globe and circle graticules. For example if sued in conjunction with NG2 the overall size range = 128:1. The circles increase by 2. Note: Both graticules would have to be used with the same microscope, eyepiece and objective.
Size: 19mm
Divisions: Grid length 4 mm
Surface: Glass sandwich
Catalog No. 50-258-82 Supplier Electron Microscopy Sciences Supplier No. 6803319
Designed to extend the sizing range of globe and circle graticules. For example if sued in conjunction with NG2 the overall size range = 128:1. The circles increase by 2. Note: Both graticules would have to be used with the same microscope, eyepiece and objective.
Size: 19mm
Divisions: Grid length 4 mm
Surface: Glass sandwich