Learn More
Medchemexpress LLC FAK inhibitor 2 | 2354405-14-2 | 98.3% | 50 MG
SDP
Catalog No. 5000338283
Encompass_Preferred
FAK inhibitor 2 is a potent focal adhesion kinase (FAK) inhibitor with an IC50 of 0.07 nM, demonstrating antitumor and anti-angiogenesis activities. This product is intended for research use only.
  • Potent FAK inhibitor (IC50 of 0.07 nM).
  • Exhibits antitumor and anti-angiogenesis activities.
  • High purity of 98.26%.
  • Soluble in DMSO at 100 mg/mL.
  • Available in various quantities.
  • Comprehensive documentation included.
  • Cited in multiple research publications.
  • Specific storage conditions for optimal stability.
  • Relevant for research in Cancer, Metabolic Disease, and more.
Catalog No. 50-003-38283 Supplier Medchemexpress LLC Supplier No. HY12858050MG
FAK inhibitor 2 is a potent focal adhesion kinase (FAK) inhibitor with an IC50 of 0.07 nM, demonstrating antitumor and anti-angiogenesis activities. This product is intended for research use only.
  • Potent FAK inhibitor (IC50 of 0.07 nM).
  • Exhibits antitumor and anti-angiogenesis activities.
  • High purity of 98.26%.
  • Soluble in DMSO at 100 mg/mL.
  • Available in various quantities.
  • Comprehensive documentation included.
  • Cited in multiple research publications.
  • Specific storage conditions for optimal stability.
  • Relevant for research in Cancer, Metabolic Disease, and more.