Learn More
Cayman Chemical FAK Inhibitor 14, Purity: ≥95%, CAS: 4506-66-5, 10 mg
SDP
Catalog No. 502972298
Encompass_Preferred
A direct inhibitor of FAK1 autophosphorylation, blocking phosphorylation of Y397 (IC50 = 1 μM); no known significant effect on the activity of a range of other kinases; promotes cell detachment and inhibits cell adhesion of cells in culture; blocks tumor growth in vivo
Catalog No. 50-297-2298 Supplier Cayman Chemical Supplier No. 1448510 MG
A direct inhibitor of FAK1 autophosphorylation, blocking phosphorylation of Y397 (IC50 = 1 μM); no known significant effect on the activity of a range of other kinases; promotes cell detachment and inhibits cell adhesion of cells in culture; blocks tumor growth in vivo