Learn More
Medchemexpress LLC FAK ligand-Linker Conjugate 1 | 2307461-45-4 | 98.9% | 583.58 | 1 ML
SDP
Catalog No. 5000403884
Encompass_Preferred
FAK ligand-Linker Conjugate 1 incorporates a ligand for FAK and a PROTAC linker, which recruit E3 ligases (such as VHL, CRBN, MDM2, and IAP). It can be extensively used for PROTAC-mediated protein degradation.
  • Incorporates a ligand for FAK.
  • Includes a PROTAC linker.
  • Recruits E3 ligases (VHL, CRBN, MDM2, IAP).
  • Used for PROTAC-mediated protein degradation.
  • For research use only.
Catalog No. 50-004-03884 Supplier Medchemexpress LLC Supplier No. HY4414810MM/1ML
FAK ligand-Linker Conjugate 1 incorporates a ligand for FAK and a PROTAC linker, which recruit E3 ligases (such as VHL, CRBN, MDM2, and IAP). It can be extensively used for PROTAC-mediated protein degradation.
  • Incorporates a ligand for FAK.
  • Includes a PROTAC linker.
  • Recruits E3 ligases (VHL, CRBN, MDM2, IAP).
  • Used for PROTAC-mediated protein degradation.
  • For research use only.