Learn More
Cell Signaling Technology FAK Monoclonal Antibody, Rabbit, 100 μg
SDP
Catalog No. NC3880687
Encompass
The FAK (D2R2E) Rabbit Monoclonal Antibody is a carrier-free antibody designed to recognize endogenous levels of total focal adhesion kinase (FAK) protein. This cytoplasmic protein tyrosine kinase is essential for integrin-mediated signal transduction, playing a critical role in controlling biological processes such as cell spreading, migration, and survival.
  • Validated for Western blotting and ELISA applications.
  • Formulated without bovine serum albumin (BSA) and azide, making it ideal for labeling techniques requiring specialized chemistry, including fluorophores, metals, lanthanides, and oligonucleotides.
  • Recombinant monoclonal antibody generated by immunizing animals with a synthetic peptide corresponding to residues surrounding Ser893 of human FAK protein.
  • Species reactivity validated for human and mouse samples.
  • Supplied in a 1X PBS buffer at a concentration of 1 mg/mL.
Catalog No. NC3880687 Supplier Cell Signaling Technology Supplier No. 87854SF
The FAK (D2R2E) Rabbit Monoclonal Antibody is a carrier-free antibody designed to recognize endogenous levels of total focal adhesion kinase (FAK) protein. This cytoplasmic protein tyrosine kinase is essential for integrin-mediated signal transduction, playing a critical role in controlling biological processes such as cell spreading, migration, and survival.
  • Validated for Western blotting and ELISA applications.
  • Formulated without bovine serum albumin (BSA) and azide, making it ideal for labeling techniques requiring specialized chemistry, including fluorophores, metals, lanthanides, and oligonucleotides.
  • Recombinant monoclonal antibody generated by immunizing animals with a synthetic peptide corresponding to residues surrounding Ser893 of human FAK protein.
  • Species reactivity validated for human and mouse samples.
  • Supplied in a 1X PBS buffer at a concentration of 1 mg/mL.