Learn More
Enzo Life Sciences Focal adhesion kinase (pTyr397) polyclonal antibody (20 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA4330020
Encompass_Preferred
Isotype: IgG. Immunogen: Synthetic peptide corresponding to a portion of FAK (focal adhesion kinase) phosphorylated at Tyr397. The sequence is conserved in mouse, rat, chicken and frog. Species reactivity: Human, Mouse Xenopus, Chicken. Specificity: Recognizes FAK phosphorylated at Tyr397. Applications: ICC, IHC, WB. Purity: Purified by antigen affinity chromatography. . Formulation: Liquid. In PBS, pH 7.4. with 50% glycerol, 1 mg/mL BSA and 0.05% sodium azide.
Catalog No. 50-201-1824