Learn More
Falcon™ 96-Well, Non-Treated, U-Shaped-Bottom Microplate
BIOPHARMAGSA_VAGREENER_CHOICE
Catalog No. 087725 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
08-772-5 Clear
1 options
This item is not returnable. View return policy
Catalog No. 08-772-5 Supplier Falcon™ Supplier No. 353910
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Color:Clear; Sterility:Non-sterile; Lid:Without Lid
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • Crystal-grade virgin polystyrene
  • Sterilized by gamma irradiation
  • Nonpyrogenic
  • Non-treated polystyrene, standard tissue culture (TC), and Corning™ PrimariaTM surface treatments available
  • Convenient, peel-open medical-style packaging for individual product presentation
  • Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
  • Falcon 96, 384, and 1536 well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Lid Without Lid
Culture Area 0.35 cm2
Array 8 x 12
Binding Type Not Treated
Color Clear
Product Type Assay Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
No. per Case 50/Cs.
No. per Pack 5/Pk.
Quantity 50/Cs.