Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Falcon™ Companion Plates for Cell Culture Inserts
BIOPHARMAGSA_VAGREENER_CHOICE

Catalog No. p-176965 Shop All Corning Products
Change view
Click to view available options
Culture Area:
3.8 cm2
9.6 cm2
Volume (Metric) Working:
1.4 to 2.3 mL
2.7 to 3.2 mL
Well Count:
6
12
Capacity (Metric):
2.3 mL (Well)
3.2 mL (Well)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Volume (Metric) Working Well Count Capacity (Metric)
08-771-22 3.8 cm2 1.4 to 2.3 mL 12 2.3 mL (Well)
08-771-24 9.6 cm2 2.7 to 3.2 mL 6 3.2 mL (Well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 08-771-22 Supplier Falcon™ Supplier No. 353503
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Feature a one-way, labyrinth lid design and condensation rings to minimize evaporation and contamination. Falcon™ Companion Plates for Cell Culture Inserts are specifically designed for use with Falcon or Corning™ BioCoat™ Cell Culture Inserts.

Falcon Cell Culture Insert Companion Plates have been specially designed for use with Falcon or Corning™ BioCoatTM Cell Culture Inserts so evaporation and contamination due to improper lid fit is eliminated.

In the Feeding Position, pipet access is improved for fluid handling on the basolateral side. Reagents can be added quickly and consistently for timed experiments. Aspiration of media from the well is easier, reducing the risk of contamination.

In the Incubation Position, Falcon Cell Culture Inserts remain locked in position in their Falcon Companion Plate wells. Media will not wick up between the insert and well wall. The unique Falcon low-evaporation lid provides a tortuous air passage system that reduces evaporation and contamination.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Closure Included Yes
Capacity (Metric) 2.3 mL (Well)
Color Clear
Culture Area 3.8 cm2
Material Polystyrene
Sterility Sterile
Surface Treatment Cell Culture
Type Multi-well Insert System
Volume (Metric) Working 1.4 to 2.3 mL
Well Count 12
For Use With (Equipment) Cell culture inserts
No. per Case 50/Cs.
No. per Pack 1/Pk.
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.