Learn More
Enterprise Technology Solutions Falcon Dust-Off Jumbo Disposable Duster, 17 oz, 2/PK
SDP
Catalog No. 5000740657
Encompass_Preferred
Jumbo-size duster removes dust and lint from computer disks, tape drives, diskettes and other electronics with pure moisture-free blasts. Formula contains a bitterant to discourage inhalant abuse. The 100 percent ozone-safe duster contains no CFCs, HCFCs, propane or butane.
Catalog No. 50-007-40657 Supplier Enterprise Technology Solutions Supplier No. FALDPSJMB2
Jumbo-size duster removes dust and lint from computer disks, tape drives, diskettes and other electronics with pure moisture-free blasts. Formula contains a bitterant to discourage inhalant abuse. The 100 percent ozone-safe duster contains no CFCs, HCFCs, propane or butane.