Learn More
Falcon™ 96-Well, Cell Culture-Treated, Flat-Bottom, Insert System Microplate
GREENER_CHOICE
Catalog No. 08771001 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
08-771-001 Clear
1 options
Catalog No. 08-771-001 Supplier Falcon™ Supplier No. 351131
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Color:Clear; Sterility:Sterile; Lid:With Lid
The Falcon 96 Multiwell Insert System is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based assays. The system has been tested for its ability to produce a differentiated monolayer of Caco-2, LLC-PK1, and MDCK cells making it an ideal platform for in vitro bioavailability and permeability studies.
This automation compatible platform is composed of a 1.0 μm pore size PET membrane-based 96 Multiwell Insert plate, a media feeder tray, and a lid. The drop-in baffle for the Feeder Tray mitigates media sloshing and lowers the risk of contamination. To analyze individual samples, simply transfer the insert plate into the Falcon 96 Square Well, Angled-Bottom Plate. If desired, the Falcon 96 Square Well, Angled-Bottom Plate may be used in conjunction with the insert for culturing the cells, eliminating the transfer step from the single-well feeder tray for sample analysis.
  • Automation compatible design
  • Format compatible with most robots and fluid handling instruments
  • Complete sample recovery
  • The Falcon 96 Square Well, Angled-Bottom Plate features an angled bottom for more complete sample utilization
  • Excellent reproducibility
  • One-piece feeder tray enhances consistency in well-to-well monolayer growth
  • Total assay flexibility—ideal for transport studies
  • System can be used with many cell lines including Caco-2, MDCK, and LLC-PK1, for basal to apical or apical to basal measurements of drug transport
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid With Lid
Culture Area 0.08 cm2
Membrane PET
Array 8 x 12
Pore Size 1 μm
Color Clear
No. of Wells 96
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
Includes Insert plate with feeder tray and lid
No. per Case 5/Cs.
No. per Pack 5/Pk.
Quantity 5/Cs.
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov