Learn More
Falcon™ Multiwell Library Storage Plate
GREENER_CHOICE
Catalog No. 08789 Shop All Corning Products
Change view
Click to view available options
No. per Pack:
25/Pk.
Catalog No. No. per Pack
08-789 25/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 08-789 Supplier Falcon™ Supplier No. 351190
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Ideal for storing, archiving, and transporting of pharmaceutical drug candidates
Falcon™ 96-well Polypropylene Microplate, not treated, round bottom, 25/pack, 50/case : The Falcon 96 well Polypropylene Library Storage Plate is convenient for compound storage : Notch H12
  • Exceedingly rigid and flat plate for consistent automated handling
  • Plate withstands repeated freeze-thaws down to -20°C
  • Low retention polypropylene and round-well design give low residual volumes after pipetting (<0.1 μL)
  • Raised well edges and flatness for heat or adhesive sealing
  • In addition to the round-bottom Falcon 96 well Library Storage Plate, V-bottom formats are also available.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Color Clear
    Material Polypropylene
    Sterility Non-sterile
    No. of Wells 96
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Product Type Storage Microplate
    Well Shape Round