Learn More
Falcon™ Pipet Controller Hydrophobic PTFE Filters
GREENER_CHOICE
Catalog No. p-4687565 Shop All Corning Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
05-100-503 0.45 μm
05-100-502 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 05-100-503 Supplier Falcon™ Supplier No. 357473
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Replacement hydrophobic PTFE disc filters
Additional filters are available as standard accessories. The filters, pipet holder, and nose piece are autoclavable.
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE
Pore Size 0.45 μm
For Use With (Equipment) For use with pipet controller
Quantity 5/Cs.