Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Falcon Safety Dust-Off™ Classic Duster

Catalog No. 1899964 Shop All Falcon Safety Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Product Type:
Classic Duster
Refill for Classic Duster
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
18-999-64 Classic Duster
18-999-65 Refill for Classic Duster
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Keep away from heat, open flames, or other sources of ignition.
Catalog No. 18-999-64 Supplier Falcon Safety Supplier No. FGS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Keep away from heat, open flames, or other sources of ignition.

Removes dust and lint from computers, electronics, cameras, keyboards, workshops and more

  • Chrome valve for dependable use
  • 100% ozone safe
  • Contains bitterant to discourage inhalant abuse
  • 10 oz. can

Specifications

Product Type Classic Duster
For Use With (Application) Removes dust and lint from computers, electronics, cameras, keyboards, workshops
Volume (English) 10 oz.
Volume (Metric) 0.29 L

Use good industrial hygiene practices in handling this material. Avoid contact with eyes and skin. Avoid breathing mists or aerosols of this product. Use only with adequate ventilation. Flammable gas may catch fire. Contact with liquid may cause frostbite. Excessive intentional inhalation may cause respiratory tract irritation and central nervous system effects (headache, dizziness). Vapors may cause dizziness or suffocation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.