Learn More
Honeywell Miller™ Falcon™ Stainless Steel Cable Self-Retracting Lifeline
Catalog No. 19085589
Change view
Click to view available options
Single
Catalog No.
19-085-589 Single
1 options
Catalog No. 19-085-589 Supplier Honeywell Miller™ Supplier No. MP20SSZ7/20FT
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For connecting a worker's full-body safety harness to an anchor point on a building or structure. SRLs prevent workers from free-falling and help to prevent injuries in the event of a fall.
  • Corrosion-resistant stainless steel and aluminum internal components extend service life and reduce replacement costs
  • Lightest composite unit on the market – easy to handle and transport to/from jobsites
  • Unique side payout design reduces wear on the entry guide bushing and cable for smooth operation with less drag
  • Engineered spring retraction tension is designed not to pull or drag on a worker to improve productivity and reduce fatigue
  • Dual swivels D3814 (unit top and snap hook) included on all cable models to minimize lifeline from twisting during use
  • Smooth, quiet operation
TRUSTED_SUSTAINABILITY
Specifications
Leg Configuration Single
Product Type Self-Retracting Lifeline (SRL)
Certifications/Compliance ANSI A10.32, ANSI Z359.14 Class B, CSA Z259.22-98, OSHA 1926.502
Diameter (English) 3/16 in.
Connector Types Steel Snap Hook, Steel Carabiner
Capacity (English) 310 lb.
Diameter (Metric) 0.48 mm
Length (English) 20 ft.
Color Silver