Learn More
Falcon™ TC-treated Multiwell Plate
GREENER_CHOICE
Catalog No. 08774183 Shop All Corning Products
Change view
Click to view available options
Well Count:
6
Catalog No. Well Count
08-774-183 6
1 options
Catalog No. 08-774-183 Supplier Falcon™ Supplier No. 355467
Add to Cart
Add to Cart
Accommodate the large media volumes required when culturing cells on microporous membranes
Falcon Cell Culture Insert Companion Plates have been specially designed for use with Falcon or Corning™ BioCoatTM Cell Culture Inserts so evaporation and contamination due to improper lid fit is eliminated.
In the Feeding Position, pipet access is improved for fluid handling on the basolateral side. Reagents can be added quickly and consistently for timed experiments. Aspiration of media from the well is easier, reducing the risk of contamination.
In the Incubation Position, Falcon Cell Culture Inserts remain locked in position in their Falcon Companion Plate wells. Media will not wick up between the insert and well wall. The unique Falcon low-evaporation lid provides a tortuous air passage system that reduces evaporation and contamination.
TRUSTED_SUSTAINABILITY
Specifications
Bottom Shape Flat
Color Clear
Material Polystyrene
Sterility Sterile
Surface Treatment Cell Culture
Type Multi-well Cell Culture Plate
Well Count 6
No. per Case 4/Cs.
No. per Pack 1/Pk.