Learn More
Chemglass Life Sciences Wire Tube Rack, HDPE-Coated, 15-Place for 15 mL Centrifuge Tubes, Blue
SDP
Catalog No. NC2323629
Encompass
Coated wire tube rack designed to hold and organize 15 mL centrifuge tubes for sample storage, transport, and routine laboratory handling. The HDPE-coated wire construction provides chemical resistance and durability; the rack is autoclavable and suitable for storage to -40° C.
  • Holds fifteen 15 mL centrifuge tubes.
  • HDPE-coated wire construction for chemical resistance.
  • Autoclavable and usable down to -40° C.
  • Slotted bottom and square wells keep tubes secure.
  • Compact, bench-top footprint for easy storage and handling.
Catalog No. NC2323629 Supplier Chemglass Life Sciences Supplier No. CLS4796001
Coated wire tube rack designed to hold and organize 15 mL centrifuge tubes for sample storage, transport, and routine laboratory handling. The HDPE-coated wire construction provides chemical resistance and durability; the rack is autoclavable and suitable for storage to -40° C.
  • Holds fifteen 15 mL centrifuge tubes.
  • HDPE-coated wire construction for chemical resistance.
  • Autoclavable and usable down to -40° C.
  • Slotted bottom and square wells keep tubes secure.
  • Compact, bench-top footprint for easy storage and handling.