Learn More
Chemglass Life Sciences Coated wire tube rack, 8-place for 50 mL centrifuge tubes (blue)
SDP
Catalog No. NC2323630
Encompass
Coated wire tube rack designed to hold eight 50 mL centrifuge tubes for transport, handling, and storage. HDPE-coated wire construction provides chemical resistance and durability; square wells and three deck levels secure tubes while a slotted bottom allows drainage. The rack is autoclavable and suitable for use down to -40°C.
  • HDPE-coated wire construction for chemical resistance.
  • Holds eight 50 mL centrifuge tubes.
  • Square wells with three deck levels secure tubes.
  • Slotted bottom allows drainage and airflow.
  • Autoclavable and rated for use to -40°C.
Catalog No. NC2323630 Supplier Chemglass Life Sciences Supplier No. CLS4796002
Coated wire tube rack designed to hold eight 50 mL centrifuge tubes for transport, handling, and storage. HDPE-coated wire construction provides chemical resistance and durability; square wells and three deck levels secure tubes while a slotted bottom allows drainage. The rack is autoclavable and suitable for use down to -40°C.
  • HDPE-coated wire construction for chemical resistance.
  • Holds eight 50 mL centrifuge tubes.
  • Square wells with three deck levels secure tubes.
  • Slotted bottom allows drainage and airflow.
  • Autoclavable and rated for use to -40°C.