Learn More
Cayman Chemical FALGPA (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502917919
Encompass_Preferred
A colorimetric collagenase substrate; selective for collagenase over trypsin, thermolysin, and elastase; cleavage of the LG peptide sequence releases the furanacryloyl group, which can be quantified by colorimetric detection at 324-345 nm as a measure of collagenase activity; has been used to identify inhibitors of C. histolyticum collagenase
Catalog No. 50-291-7919 Supplier Cayman Chemical Supplier No. 3737410 MG
A colorimetric collagenase substrate; selective for collagenase over trypsin, thermolysin, and elastase; cleavage of the LG peptide sequence releases the furanacryloyl group, which can be quantified by colorimetric detection at 324-345 nm as a measure of collagenase activity; has been used to identify inhibitors of C. histolyticum collagenase