Learn More
Anton Paar Falling ball viscometer with adjustable angle for different viscosities. Measures intrinsic viscosity, K-value, and molar mass. Referenced in USP 913 and Ph. Eur. 2.2.49. Automated filling/cleaning and optional combination with refractive index and pH.