Learn More
Medchemexpress LLC Faltan | 133-07-3 | >98.2% | 500 MG
SDP
Catalog No. 5000409638
Encompass_Preferred
Faltan is a dicarboximide fungicide widely used on vines and several vegetable crops. It also exhibits a cytotoxic effect on human bronchial epithelial cells.
  • Dicarboximide fungicide
  • Used on vines and various vegetable crops
  • Cytotoxic effect on human bronchial epithelial cells
Catalog No. 50-004-09638 Supplier Medchemexpress LLC Supplier No. HYB1878500MG
Faltan is a dicarboximide fungicide widely used on vines and several vegetable crops. It also exhibits a cytotoxic effect on human bronchial epithelial cells.
  • Dicarboximide fungicide
  • Used on vines and various vegetable crops
  • Cytotoxic effect on human bronchial epithelial cells