Learn More
CPC Scientific FAM-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502108598
Encompass
Pituitary hormone N-terminal synthetic fragment that stimulates the corticosteroid synthesis and secretion in the adrenal cortex. This peptide is more active than ACTH in terms of corticosteroid releasing activity in vitro.ONE-LETTER SEQUENCE: Fam-SYSMEHFRWGKPVGKKRRPVKVYPMOLECULAR FORMULA: C157H220N40O37S1MOLECULAR WEIGHT:3291.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [16960-16-0], RESEARCH AREA: HormonalREFERENCES: Y.F.Jacquet, Science, 201, 1032 (1978)
Catalog No. 50-210-8598 Supplier CPC Scientific Supplier No. ACTH004A
Pituitary hormone N-terminal synthetic fragment that stimulates the corticosteroid synthesis and secretion in the adrenal cortex. This peptide is more active than ACTH in terms of corticosteroid releasing activity in vitro.ONE-LETTER SEQUENCE: Fam-SYSMEHFRWGKPVGKKRRPVKVYPMOLECULAR FORMULA: C157H220N40O37S1MOLECULAR WEIGHT:3291.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [16960-16-0], RESEARCH AREA: HormonalREFERENCES: Y.F.Jacquet, Science, 201, 1032 (1978)