Learn More
Medchemexpress LLC FAM amine, 6-isomer | 2183440-42-6 | MFCD31580128 | 98.1% | 510.97 g/mol | C27H27ClN2O6 | 10 MG
SDP
Catalog No. 5000297447
Encompass_Preferred
FAM amine, 6-isomer is a fluorescein-derived fluorescent dye bearing a primary amine at the 6-position. It provides a bright fluorescein signal for labeling oligonucleotides, peptides, and proteins in fluorescence assays and imaging. Supplied as a small-mass research reagent, it enables covalent conjugation through amine-reactive chemistry.
  • Fluorescent dye with primary amine for direct conjugation
  • Pure 6-isomer for consistent spectral and labeling performance
  • Molecular formula C27H27ClN2O6; molecular weight 510.97 g/mol
  • High purity suitable for research applications
  • Available in small milligram quantities for laboratory use
Catalog No. 50-002-97447 Supplier Medchemexpress LLC Supplier No. HYD133610MG
FAM amine, 6-isomer is a fluorescein-derived fluorescent dye bearing a primary amine at the 6-position. It provides a bright fluorescein signal for labeling oligonucleotides, peptides, and proteins in fluorescence assays and imaging. Supplied as a small-mass research reagent, it enables covalent conjugation through amine-reactive chemistry.
  • Fluorescent dye with primary amine for direct conjugation
  • Pure 6-isomer for consistent spectral and labeling performance
  • Molecular formula C27H27ClN2O6; molecular weight 510.97 g/mol
  • High purity suitable for research applications
  • Available in small milligram quantities for laboratory use