Learn More
Apexbio Technology LLC FAM azide, 5- isomer, 5mg. Cas: MFCD: NA. Fluorescent lablels
SDP
Supplier:  Apexbio Technology LLC A81175
Encompass_Preferred
FAM azide for Click chemistry labeling. FAM remains one of the most popular fluorescent lablels for various application. Most instruments capable of fluorescence detection, ranging from plate readers to fluorescence microscopes, are able to work in FAM channel. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5019